Hand Sanitizer, Mortech
Catalog # 76549-674
Supplier: Mortech Manufacturing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Capacity:16 oz.
- Cat. no.:76549-674
Specifications
About this item
Alcohol content 70% hand sanitizing gel, kills viruses and bacteria, highly effective for commercial and breakroom/office surface cleaning.
- Topical solution
- Alcohol antiseptic 70%
- Made in the USA
- Liquid product (not gel)