Body Bag, Guardian Safe View, Poly, Envelope Zipper, No Handles, Mortech
Catalog # 76549-666
Supplier: Mortech Manufacturing
Specifications
- Dimensions:96Lx36"W
- Cat. no.:76549-666
Specifications
About this item
Unique optically clear interior panel on the Mopec BE175 allows viewing the remains contained in the bag with minimal exposure risk to personnel. Both panels are secured with envelope style heavy-duty zippers with dual zipper pulls for easy access.
- Standard duty, 10 Mil reinforced coextruded polyethylene
- Envelope (C-shape) zipper
- Heat sealed seams
- Weight capacity 300 lbs.
- Suitable for cremation
- Clear viewing panel