Body Bag, Envelope Zipper, CP Heavy Duty (1000 lbs.), Polyethylene with Reinforced Nylon, Mortech
Catalogue # 76549-656
Fournisseur: Mortech Manufacturing
Spécifications
- Dimensions:40W×90"L
- Cat. no.:76549-656
Spécifications
À propos de cet article
Heavy duty body bag with an envelope style zipper, no handles, and available in white.
- 8 mil high strength, white or black, polyethylene with reinforced nylon
- Envelope style, rust resistant zipper
- No handles
- Shelf life 10 years
- Recommended for cremation