Mopec Safe Seal System, Replacement Roll, Mortech
Mopec Safe Seal System, 150′ replacement roll
Catalog # 76549-652
Supplier: Mopec
Supplier #: BE162
Jump to:
Product Details & Documents
Made of a military tested flexible material that meets Biosafety Level 4 (BSL-4) containment standards and is a fully hermetic body bag for infectious human remains. It meets IATA requirements for air transport and repatriation. Mopec Safe Seal is safe for cremation.
- 49 year shelf life
- 150' long
- 8 mil
- Each roll only needs 8×8" of required storage space
- Safe for cremation
Specifications
- Désignation:Mopec Safe Seal System, 150′ replacement roll