Instrument Trays and Covers, Mortech
Fournisseur: Mortech Manufacturing
Certificats
À propos de cet article
Instrument trays and covers are made of stainless steel and can be used for a number of facility needs.
- All stainless steel, seamless construction
- Coved corners for easy cleaning
- Covers have recessed strap handles for ease in handling and storing
Availabe in a variety of sizes. Cover sold separately.
Frequently Bought Together