Drain Bucket, Plastic, Mortech
Catalog # 76549-552
Supplier: Mortech Manufacturing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Capacity:14 qt.
- Color:Gray
- Description:Drain bucket, plastic
- Cat. no.:76549-552
Specifications
About this item
This quality utility bucket and kickstand can be used for a variety of facility needs.
- Graduated in pints, quarts, gallons, and liters
- Meets FDA standards
- 14 quart total capacity
This injection-molded plastic bucket is designed to resist most acids, alkalies, paint, chemicals, and cleaning compounds.