Caliper, Analog, Mortech
3.75" (95 mm)
Catalog # 76549-516
Supplier: Mopec
Supplier #: BA040
Jump to:
Product Details & Documents
Excellent for measuring all tissue specimens with a sliding caliper and a finger tip nut to lock down ruler. Tool has graduated etched standard and metric markings.
- Opens to 3" (76 mm)
- Premium grade stainless steel
Specifications
- Longueur:3.75" (95 mm)