Scalpel Blades, No. 22, Nonsterile, Bulk, Mortech
Catalogue # 76549-460
Fournisseur: Mortech Manufacturing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Cat. no.:76549-460
Spécifications
À propos de cet article
No. 22 Blades for all post-mortem dissection procedures. Buy in bulk for convenience and savings.
- Nonsterile stainless steel
- Pack of 500
- Fits handles AJ101, AJ102 and AJ126