ContactUsLinkComponent
Scalpel Handle For Double Edged Blade, Mortech
Catalogue # 76549-432
Fournisseur:  Mortech Manufacturing
undefined
Scalpel Handle For Double Edged Blade, Mortech
Catalogue # 76549-432
Fournisseur:  Mortech Manufacturing
Supplier Number:  AJ127
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Scalpel handle for double edged blade
  • Cat. no.:
    76549-432
Spécifications
À propos de cet article
Mopec’s double edge blade handle is ideal for trimming specimens and is a versatile tool for any laboratory.
  • Compatible with Mopec AJ111 double edge blades
  • Blades not included
The double edge blade is securely held in place, yet easy to replace. The weighted pencil-style handle provides a firm, mobile grip.