ContactUsLinkComponent
Scalpel Handle, Molded Grip, No. 6, Mortech
Catalogue # 76549-430
Fournisseur:  Mopec
undefined
Scalpel Handle, Molded Grip, No. 6, Mortech
Catalogue # 76549-430
Fournisseur:  Mopec
Supplier Number:  AJ126
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Length:
    5.5"
  • Cat. no.:
    76549-430
Spécifications
À propos de cet article
This molded handle has a smooth grip for comfortable post-mortem dissection. Fits blade sizes #20 – #23.
  • Premium grade stainless steel with ergonomic plastic handle
  • Fits #20 – #23 blades
It can easily be cleaned and autoclaved for long-lasting life.