Blade Remover, Disposable, Mortech
Blade remover, disposable
Catalog # 76549-414
Supplier:   Mopec
 :  AJ116
Blade Remover, Disposable, Mortech
Catalog # 76549-414
Supplier:  Mopec
Blade Remover, Disposable, Mortech
Blade Remover, Disposable, Mortech
Blade remover, disposable
Price
$185.15
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Designed to safely remove dull contaminated blade from scalpel handle.
  • Pack of 10
  • Autoclavable
  • Holds 100 blades each
  • Not compatible with double sided blades
Simply insert and press handle to dispense old blade safely into sealed container.
Spécifications
  • Désignation:
    Blade remover, disposable