ContactUsLinkComponent
Scalpel Handle, Bard Parker, No. 8, Mortech
Scalpel Handle, Bard Parker, No. 8, Mortech
Catalog # 76549-412
Supplier:  Mopec
undefined
Scalpel Handle, Bard Parker, No. 8, Mortech
Catalog # 76549-412
Supplier:  Mopec
Supplier Number:  AJ115
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Length:
    6.5"
  • Cat. no.:
    76549-412
Specifications
About this item
Handle for #60 and #70 blades. Durable and reusable.
  • Smooth, ergonomic plastic handles
  • Fits #60 and #70 blades
  • Latex free