Chalk Calvaria Elevator, Mortech | Avantor
Chalk Calvaria Elevator, Mortech
Chalk calvaria elevator
Catalog # 76549-380
Supplier:   Mopec
Fournisseur #:  AH200
Chalk Calvaria Elevator, Mortech
Catalog # 76549-380
Supplier:  Mopec
Chalk Calvaria Elevator, Mortech
Chalk Calvaria Elevator, Mortech
Chalk calvaria elevator
Price
$616.03
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Chalk Calvaria Elevator is designed with both function and comfort in mind. The penetrating edge is only 10 mm long which prevents over penetration of the cranial cavity and laceration to the brain. The heavy-duty strike plate and handle are mounted at a right angle to the penetrating edge. This makes the use of a mallet easier while holding the tool and allows the user to apply torque to remove the calvaria.
  • Stainless steel
With the Chalk Calvaria Elevator any discomfort from the torque method is eliminated, which in turn will reduce user injury.
Spécifications
  • Désignation:
    Chalk calvaria elevator