Scissors Sharpener, Electric Diamond Hone, Mortech | Avantor
Scissors Sharpener, Electric Diamond Hone, Mortech
Scissors sharpener, electric diamond hone
Catalog # 76549-374
Supplier:   Mopec
Fournisseur #:  AH118
Scissors Sharpener, Electric Diamond Hone, Mortech
Catalog # 76549-374
Supplier:  Mopec
Scissors Sharpener, Electric Diamond Hone, Mortech
Scissors Sharpener, Electric Diamond Hone, Mortech
Scissors sharpener, electric diamond hone
Price
$471.41
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A revolutionary development in scissor sharpening. Produces quality sharpness with unparalleled simplicity. For incredibly sharp, straight scissors. Sharpens from tip to handle without the need to disassemble scissor. 100% diamond abrasive in stages 1 and 2 for superior results.
  • Two (2) stages, sharpening and honing, for a razor sharp, longer lasting double bevel edge
  • Precise bevel angle control for a 20° edge
  • Ready to use, left or right handed
  • Safe for all quality scissors
  • 120 V
Double bevel edge stays sharp longer. Never-tempered patented technology eliminates guesswork for sharper edges. Compact, easy, safe, and rugged design.
Ordering information: 1-year manufacturers warranty.
Spécifications
  • Désignation:
    Scissors sharpener, electric diamond hone