Knife Sharpener, Variable Speed, Mortech
Knife Sharpener, Variable Speed, Mortech
 :   Mopec
Knife Sharpener, Variable Speed, Mortech
 :  Mopec
Knife Sharpener, Variable Speed, Mortech
Knife Sharpener, Variable Speed, Mortech
This variable speed sharpener creates a precise angle and a consistent, exact bevel by sharpening, thinning, and honing which results in a stronger cutting edge. The honing wheels are mounted on parallel shafts that rotate in opposite directions.
  • Stainless steel case
Available in US and International voltage.
Ordering information: Replacement honing wheels available (See model AH034).
Product Family Options
  • Désignation
  • 110 V
    Each
    Each - $4,894.32
    $4,894.32
    Each
       $4,894.32
     : AH032
  • 220 V
    Each
    Each - $5,313.41
    $5,313.41
    Each
       $5,313.41
     : AH033