Knife Sharpener, 3 Step, Mortech | Avantor
Knife Sharpener, 3 Step, Mortech
Knife Sharpener, 3 Step, 12.5"
Catalog # 76549-328
Supplier:   Mopec
Fournisseur #:  AH030
Knife Sharpener, 3 Step, Mortech
Catalog # 76549-328
Supplier:  Mopec
Knife Sharpener, 3 Step, Mortech
Knife Sharpener, 3 Step, Mortech
Knife Sharpener, 3 Step, 12.5"
Price
$528.72
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mopec’s three-step knife sharpener is constructed of durable polypropylene.
  • Includes three changeable ceramic rods and complete instructions
Sharpen virtually any dull knife with the ceramic rods standing vertically or laying horizontally on your bench top. The AH030 can be used for dull or worn edges, partial dull edges and quick touch ups.
Requires no oil or water. Rods can be rotated for sharp new surfaces providing longer stone life.
Spécifications
  • Désignation:
    Knife Sharpener, 3 Step
  • Longueur:
    12.5"