Bone Rongeurs, Blumenthal Angled, Mortech | Avantor
Bone Rongeurs, Blumenthal Angled, Mortech
Supplier:   Mopec
Bone Rongeurs, Blumenthal Angled, Mortech
Supplier:  Mopec
Certificates
Bone Rongeurs, Blumenthal Angled, Mortech
Bone Rongeurs, Blumenthal Angled, Mortech
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Perfect instrument for removing small bone fragments during post-mortem dissection. Variety of angles from which to choose.
  • German grade satin
Available in various angles.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Availability
    Price
  • 30 Degrees
    6"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $107.12
    $107.12
    Each
       $107.12
    Supplier Number: AG030
  • 45 Degrees
    6"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $107.12
    $107.12
    Each
       $107.12
    Supplier Number: AG031
  • 90 Degrees
    6"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $112.15
    $112.15
    Each
       $112.15
    Supplier Number: AG032