Bone Chisel, Brunetti’s, Mortech | Avantor
Bone Chisel, Brunetti’s, Mortech
Supplier:   Mopec
Bone Chisel, Brunetti’s, Mortech
Supplier:  Mopec
Certificates
Bone Chisel, Brunetti’s, Mortech
Bone Chisel, Brunetti’s, Mortech
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Brunetti's chisels are specifically designed to score, cut, scrape, clean and sculpt around the spinal cord.
  • Standard grade stainless steel
Available in right handed and left handed options.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Availability
    Price
  • Left-hand handle
    11"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $169.46
    $169.46
    Each
       $169.46
    Supplier Number: AG016
  • Right-hand handle
    11"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $299.22
    $299.22
    Each
       $299.22
    Supplier Number: AG015