Bone Chisel, Lambotte, Straight, Mortech
Bone Chisel, Lambotte, Straight, Mortech
Supplier:   Mopec
Bone Chisel, Lambotte, Straight, Mortech
Supplier:  Mopec
Certificates
Bone Chisel, Lambotte, Straight, Mortech
Bone Chisel, Lambotte, Straight, Mortech
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Osteotomes, also called bone chisels, are used to score, cut, scrape, clean and sculpt bone specimen.
  • Stainless steel
  • Lambotte pattern
Available in standard or premium grade with various edges.
Product Family Options
Informations sur le produit
  • Grade
    Longueur
    Tip Dimensions
    Availability
    Price
  • Standard
    8"
    0.5"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $107.12
    $107.12
    Each
       $107.12
    Supplier Number: AG005
  • Standard
    8"
    1"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $112.15
    $112.15
    Each
       $112.15
    Supplier Number: AG006
  • Premium
    9"
    0.5"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $364.08
    $364.08
    Each
       $364.08
    Supplier Number: AG105
  • Premium
    9"
    1"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $411.44
    $411.44
    Each
       $411.44
    Supplier Number: AG106