Rib Shears, Plain Curved, Mortech
9"
  76549-172
 :   Mopec
 :  AF002
Rib Shears, Plain Curved, Mortech
  76549-172
 :  Mopec
Rib Shears, Plain Curved, Mortech
Rib Shears, Plain Curved, Mortech
9"
$391.51
Each
Rib shears with curved end and semi-box eccentric lock to hold and cut through bone efficiently and safely.
  • Standard grade stainless steel
  • Longueur:
    9"