Dissection Tool, Light Duty, Mortech
15.5 cm (6⅛")
Catalog # 76549-122
Supplier:   Mopec
 :  AD120
Dissection Tool, Light Duty, Mortech
Catalog # 76549-122
Supplier:  Mopec
Dissection Tool, Light Duty, Mortech
Dissection Tool, Light Duty, Mortech
15.5 cm (6⅛")
Price
$26.96
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ideal for delicate and light duty post-mortem dissection procedures.
  • Dual prong, sharp narrow
Available in a variety of thin to wide spaced curved prongs for gently isolating and retracting fascia, ligament tissue, nerve isolation, nerve teasing and spinal cord dissection. Serrated total perimeter grip assures secure tissue handling.
Specifications
  • Longueur:
    15.5 cm (6⅛")