Retractor, Blunt/Sharp, Mortech
 :   Mopec
Retractor, Blunt/Sharp, Mortech
 :  Mopec
Retractor, Blunt/Sharp, Mortech
Retractor, Blunt/Sharp, Mortech
High quality retractor assists in separating tissues during autopsy procedures to allow for deeper cutting or probing.
  • Stainless steel
  • 3×4 prongs
Available in multiple lengths with sharp or blunt prongs.
Product Family Options
  • Longueur
    Tip Configuration
  • 14.0 cm (5½")
    Blunt
    Each
    Each - $122.13
    $122.13
    Each
       $122.13
     : AD048
  • 14.0 cm (5½")
    Sharp
    Each
    Each - $127.14
    $127.14
    Each
       $127.14
     : AD047
  • 24.1 cm (9½")
    Blunt
    Each
    Each - $162.01
    $162.01
    Each
       $162.01
     : AD050
  • 24.1 cm (9½")
    Sharp
    Each
    Each - $162.01
    $162.01
    Each
       $162.01
     : AD049