Stapler, Postmortem, Mortech | Avantor
Stapler, Postmortem, Mortech
Stapler, postmortem
Catalog # 76549-070
Supplier:   Mopec
Fournisseur #:  AD046
Stapler, Postmortem, Mortech
Catalog # 76549-070
Supplier:  Mopec
Stapler, Postmortem, Mortech
Stapler, Postmortem, Mortech
Stapler, postmortem
Price
$154.50
Log in pour la disponibilité
Each
Product Details & Documents
Postmortem stapler provides a safe, fast and efficient option for skin closure.
  • Staples 0.25"
  • Pre-loaded with 35 staples
  • Disposable
Reduces needle and sharps exposures. Arrow guide assists in accurate staple placement. It also features an easy-to-read staple counter.
Spécifications
  • Désignation:
    Stapler, postmortem