Section Lifter, Double Ended, Mortech
15.2 cm (6")
Catalog # 76549-062
Supplier: Mopec
Fournisseur #: AD040
Jump to:
Product Details & Documents
Double ended section lifter to separate and lift tissue during post-mortem procedures.
- Standard grade stainless steel
Spécifications
- Longueur:15.2 cm (6")