Retractor/Lifter, Sharp/Blunt, Curved, Mortech
Supplier:   Mopec
Retractor/Lifter, Sharp/Blunt, Curved, Mortech
Supplier:  Mopec
Certificates
Retractor/Lifter, Sharp/Blunt, Curved, Mortech
Retractor/Lifter, Sharp/Blunt, Curved, Mortech
2 Options Available Below
Product Details & Documents
Paddle end ideal for isolating and retracting tissue during dissection procedures.
  • Stainless steel
  • Three prongs
Available with sharp or blunt prongs.
Product Family Options
Informations sur le produit
  • Longueur
    Curvature
    Tip Configuration
    Availability
    Price
  • 16.5 cm (6½")
    Curved
    Blunt
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $259.28
    $259.28
    Each
       $259.28
    Supplier Number: AD031
  • 16.5 cm (6½")
    Curved
    Sharp
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $259.28
    $259.28
    Each
       $259.28
    Supplier Number: AD030