Probe, Pediatric Bowman, Mortech | Avantor
Probe, Pediatric Bowman, Mortech
Supplier:   Mopec
Probe, Pediatric Bowman, Mortech
Supplier:  Mopec
Certificates
Probe, Pediatric Bowman, Mortech
Probe, Pediatric Bowman, Mortech
2 Options Available Below
Product Details & Documents
Bowman probe is designed with sterling silver for use in pediatric dissection cases.
  • Sterling silver
  • Double ended
Available in multiple sizes.
Product Family Options
Informations sur le produit
  • Taille
    Longueur
    Availability
    Price
  • 00-0
    5"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $166.96
    $166.96
    Each
       $166.96
    Supplier Number: AD023
  • 000-0000
    5"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $189.46
    $189.46
    Each
       $189.46
    Supplier Number: AD024