Bullet Probe, 18", Mortech
Supplier: Mopec
Jump to:
À propos de cet article
Designed for tracking bullet path and angle of entry. Blunt probing end has a rounded tip to minimize tissue damage.
- Stainless steel
Available in multiple calibers.
3 Options Available Below
Product Details & Documents
Designed for tracking bullet path and angle of entry. Blunt probing end has a rounded tip to minimize tissue damage.
- Stainless steel
Available in multiple calibers.