Postmortem Needle Puller, Mortech
6.5"
Catalog # 76549-036
Supplier: Mopec
Fournisseur #: AD018
Jump to:
Product Details & Documents
This instrument has a curved end that holds the postmortem needle firmly allowing easy tissue pull through.
- Standard grade stainless steel
It also features a ribbed handle.
Spécifications
- Taille:6.5"