Forceps, Hemostatic, Disposable, Mortech
 :   Mopec
Forceps, Hemostatic, Disposable, Mortech
 :  Mopec
Forceps, Hemostatic, Disposable, Mortech
Forceps, Hemostatic, Disposable, Mortech
Disposable hemostatic forceps ideal for a wide variety of laboratory and post-mortem dissection procedures.
  • Stainless steel
  • Serrated tip
  • Pack of 12
Available with straight or curved edges.
Product Family Options
  • Longueur
    Curvature
  • 5"
    Straight
    Each
    Each - $94.67
    $94.67
    Each
       $94.67
     : AC601
  • 5"
    Curved
    Each
    Each - $89.66
    $89.66
    Each
       $89.66
     : AC602
  • 5.5"
    Straight
    Each
    Each - $94.67
    $94.67
    Each
       $94.67
     : AC603
  • 5.5"
    Curved
    Each
    Each - $94.67
    $94.67
    Each
       $94.67
     : AC604