Forceps, Extracting, #150A, Mortech | Avantor
Forceps, Extracting, #150A, Mortech
7.125"
Catalog # 76548-872
Supplier:   Mopec
Fournisseur #:  AB303
Forceps, Extracting, #150A, Mortech
Catalog # 76548-872
Supplier:  Mopec
Forceps, Extracting, #150A, Mortech
Forceps, Extracting, #150A, Mortech
7.125"
Price
$107.12
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Extracting forceps are used for post-mortem dental removal on upper incisors, cuspids, bicuspids and roots.
Spécifications
  • Longueur:
    7.125"