Forceps, Extracting, #32, Mortech | Avantor
Forceps, Extracting, #32, Mortech
7.25"
Catalog # 76548-868
Supplier:   Mopec
Fournisseur #:  AB301
Forceps, Extracting, #32, Mortech
Catalog # 76548-868
Supplier:  Mopec
Forceps, Extracting, #32, Mortech
Forceps, Extracting, #32, Mortech
7.25"
Price
$107.12
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Post-mortem extracting forceps used for upper cuspids, bicuspids, molars and roots.
  • German grade stainless steel
  • Universal American pattern handle
Spécifications
  • Longueur:
    7.25"