Forceps, Flat Angle Tip, Delicate, Mortech | Avantor
Forceps, Flat Angle Tip, Delicate, Mortech
3.75"
Catalog # 76548-838
Supplier:   Mopec
Fournisseur #:  AB133
Forceps, Flat Angle Tip, Delicate, Mortech
Catalog # 76548-838
Supplier:  Mopec
Forceps, Flat Angle Tip, Delicate, Mortech
Forceps, Flat Angle Tip, Delicate, Mortech
3.75"
Price
$194.46
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Raised angled flat platform tip allows for grasping the finest tissue with a firm but controlled grip.
  • Premium grade stainless steel
  • Angle tips
  • Smooth grip
Spécifications
  • Longueur:
    3.75"