Forceps, Adson Dressing, Toothed, Delicate, Mortech
Forceps, Adson Dressing, Toothed, Delicate, Mortech
4.75"
Catalog # 76548-828
Supplier:   Mopec
Supplier #:  AB128
Forceps, Adson Dressing, Toothed, Delicate, Mortech
Catalog # 76548-828
Supplier:  Mopec
Forceps, Adson Dressing, Toothed, Delicate, Mortech
Forceps, Adson Dressing, Toothed, Delicate, Mortech
4.75"
Price
152,03 $
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Adson dressing forceps are designed to hold and manipulate delicate tissues and dressing materials such as cotton or gauze during any post-mortem dissection procedure.
  • Premium grade stainless steel
  • 1×2 teeth
Extra broad ribbed grip.
Specifications
  • Longueur:
    4.75"