Forceps, Narrow Tip, Delicate, Curved, Mortech | Avantor
Forceps, Narrow Tip, Delicate, Curved, Mortech
5"
Catalog # 76548-788
Supplier:   Mopec
Fournisseur #:  AB108
Forceps, Narrow Tip, Delicate, Curved, Mortech
Catalog # 76548-788
Supplier:  Mopec
Forceps, Narrow Tip, Delicate, Curved, Mortech
Forceps, Narrow Tip, Delicate, Curved, Mortech
5"
Price
$122.13
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mopec’s standard pattern dressing forceps have serrated tips and ribbed grip to assist in delicate post-mortem dissection.
  • Premium grade stainless steel
  • Curved with delicate narrow tips
Spécifications
  • Longueur:
    5"