Forceps, Brown Adson, Mortech
Forceps, Brown Adson, Mortech
4.75"
Catalog # 76548-772
Supplier:   Mopec
Supplier #:  AB092
Forceps, Brown Adson, Mortech
Catalog # 76548-772
Supplier:  Mopec
Forceps, Brown Adson, Mortech
Forceps, Brown Adson, Mortech
4.75"
Price
$57.22
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
7×7 teeth Adson forceps. All forceps are made from stainless steel.
  • Standard grade stainless steel
  • Extra broad ribbed grip
  • Straight with 7×7 teeth tips
Specifications
  • Longueur:
    4.75"