ContactUsLinkComponent
Forceps, CutMATE Starter Kit, Mortech
Forceps, CutMATE Starter Kit, Mortech
Catalog # 76548-748
undefined
Forceps, CutMATE Starter Kit, Mortech
Catalog # 76548-748
Supplier Number:  AB070
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    CutMATE forceps starter kit
  • Cat. no.:
    76548-748
Specifications
About this item
CutMATE forceps were developed to overcome the variability of tissue specimen thickness. They will allow cutting consistent tissue specimen thickness of 2, 3, or 4 mm.
  • 304 stainless steel
The unique 'trident-like' design of the 35 mm long ergonomically angled prongs, ensures uniform tissue thickness cutting of tissue lengths that will fit standard cassettes. The inside surface of the prongs is specially textured to provide the extra grip needed for stabilizing and slicing difficult and hard to hold specimens.
Delivery: Includes (4) forceps, decontamination rack, and calibration device.