Forceps, Rubber Tipped, Mortech
Forceps, Rubber Tipped, Mortech
 :   Mopec
Forceps, Rubber Tipped, Mortech
 :  Mopec
Forceps, Rubber Tipped, Mortech
Forceps, Rubber Tipped, Mortech
Soft rubber tipped forceps allow for secure post-mortem dissection with minimal tissue damage or compression. An excellent option for techniques that require no metal exposure. Blue tips provide excellent contrast for specimen photography.
  • Standard grade stainless steel
  • Rubber rounded tips
Available in a variety of lengths.
NOTE: Do not autoclave. Rinse under water or wipe clean with any of our disinfecting cleaners.
Product Family Options
  • Longueur
  • 6"
    Each
    Each - $39.77
    $39.77
    Each
       $39.77
     : AB060
  • 8"
    Each
    Each - $54.75
    $54.75
    Each
       $54.75
     : AB061
  • 10"
    Each
    Each - $67.22
    $67.22
    Each
       $67.22
     : AB062
  • 12"
    Each
    Each - $67.22
    $67.22
    Each
       $67.22
     : AB063