Forceps, Ergonomic, Angled Tip, Mortech | Avantor
Forceps, Ergonomic, Angled Tip, Mortech
7"
Catalog # 76548-730
Supplier:   Mopec
Fournisseur #:  AB055
Forceps, Ergonomic, Angled Tip, Mortech
Catalog # 76548-730
Supplier:  Mopec
Forceps, Ergonomic, Angled Tip, Mortech
Forceps, Ergonomic, Angled Tip, Mortech
7"
Price
191,97 $
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Unique perforated paddle handles for secure slip-free grip.
  • Stainless steel
  • Broad grip
  • Angled tip
Fingertip aligned perforations offer precise maneuvering. Extremely lightweight and versatile for a wide variety of tissue specimens. Can be cleaned with any laboratory cleaner or disinfectant.
Spécifications
  • Longueur:
    7"