Forceps, Ergonomic, Mortech | Avantor
Forceps, Ergonomic, Mortech
Supplier:   Mopec
Forceps, Ergonomic, Mortech
Supplier:  Mopec
Certificates
Forceps, Ergonomic, Mortech
Forceps, Ergonomic, Mortech
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Unique perforated paddle handles for secure slip-free grip. Fingertip aligned perforations offer precise maneuvering. Extremely lightweight and versatile for a wide variety of tissue specimens. Can be cleaned with any laboratory cleaner or disinfectant.
  • Stainless steel
  • Broad grip
Available with smooth or serrated tips.
Product Family Options
Informations sur le produit
  • Longueur
    Tip Style
    Availability
    Price
  • 5.5"
    Smooth
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $191.97
    $191.97
    Each
       $191.97
    Supplier Number: AB057
  • 5.5"
    Serrated
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $191.97
    $191.97
    Each
       $191.97
    Supplier Number: AB058
  • 7"
    Smooth
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $191.97
    $191.97
    Each
       $191.97
    Supplier Number: AB054
  • 7"
    Serrated
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $191.97
    $191.97
    Each
       $191.97
    Supplier Number: AB056