Forceps, Ergonomic, Tooth Tip, Mortech | Avantor
Forceps, Ergonomic, Tooth Tip, Mortech
7"
Catalog # 76548-726
Supplier:   Mopec
Fournisseur #:  AB053
Forceps, Ergonomic, Tooth Tip, Mortech
Catalog # 76548-726
Supplier:  Mopec
Forceps, Ergonomic, Tooth Tip, Mortech
Forceps, Ergonomic, Tooth Tip, Mortech
7"
Price
$191.97
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Tooth tipped forceps with a broad ribbed handle for a secure grip. The perforations allow for an incredibly light weight instrument.
  • Stainless steel
  • Broad grip
  • Tooth tip
Spécifications
  • Longueur:
    7"