Forceps, Bayonet, Mortech | Avantor
Forceps, Bayonet, Mortech
6.5"
Catalog # 76548-722
Supplier:   Mopec
Fournisseur #:  AB050
Forceps, Bayonet, Mortech
Catalog # 76548-722
Supplier:  Mopec
Forceps, Bayonet, Mortech
Forceps, Bayonet, Mortech
6.5"
Price
$204.41
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ergonomically designed for a more comfortable wrist position during post-mortem dissections. Offers speculum function for hollow cavity specimens such as uterus, bladder and colon.
  • Standard grade stainless steel
  • Off-set serrated tip
Spécifications
  • Longueur:
    6.5"