Forceps, Dressing, Mortech
6"
  76548-710
 :   Mopec
 :  AB044
Forceps, Dressing, Mortech
  76548-710
 :  Mopec
Forceps, Dressing, Mortech
Forceps, Dressing, Mortech
6"
$44.78
Each
Dressing forceps are ideal for all post-mortem examinations and dissection procedures.
  • Standard grade stainless steel
  • Ribbed grip
  • Smooth tip
  • Longueur:
    6"