Forceps, Iris Tissue, Fine Point, Mortech
4.5"
  76548-704
 :   Mopec
 :  AB041
Forceps, Iris Tissue, Fine Point, Mortech
  76548-704
 :  Mopec
Forceps, Iris Tissue, Fine Point, Mortech
Forceps, Iris Tissue, Fine Point, Mortech
4.5"
$42.29
Each
Sharp, curved tip allows for a secure grip with minimal tissue compression during post-mortem dissection.
  • Standard grade stainless steel
  • Curved, fine point tips
  • Ribbed grip
  • Longueur:
    4.5"