Forceps with Magnifier, Mortech
Forceps with Magnifier, Mortech
4"
  76548-700
 :   Mopec
Supplier #:  AB039
Forceps with Magnifier, Mortech
  76548-700
 :  Mopec
Forceps with Magnifier, Mortech
Forceps with Magnifier, Mortech
4"
144,52 $
Each
These forceps provide 10X magnification. Very helpful with tight extractions.
  • Standard grade stainless steel
  • Serrated tips
  • Ribbed grip
  • Longueur:
    4"