Forceps, Semken Iris Tissue Dissecting, Mortech | Avantor
Forceps, Semken Iris Tissue Dissecting, Mortech
5"
Catalog # 76548-688
Supplier:   Mopec
Fournisseur #:  AB033
Forceps, Semken Iris Tissue Dissecting, Mortech
Catalog # 76548-688
Supplier:  Mopec
Forceps, Semken Iris Tissue Dissecting, Mortech
Forceps, Semken Iris Tissue Dissecting, Mortech
5"
Price
$42.29
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Sharp interlocking teeth for secure grip with minimal tissue compression.
  • Standard grade stainless steel
  • Ribbed grip
  • 1×2 teeth
Spécifications
  • Longueur:
    5"