Forceps, Adson Tissue, Mortech
Forceps, Adson Tissue, Mortech
4.75"
  76548-678
 :   Mopec
Supplier #:  AB028
Forceps, Adson Tissue, Mortech
  76548-678
 :  Mopec
Forceps, Adson Tissue, Mortech
Forceps, Adson Tissue, Mortech
4.75"
$52.25
Each
Extra broad ribbed grip for delicate, post-mortem, precision dissection.
  • Standard grade stainless steel
  • Extra broad ribbed grip
  • Straight with 1×2 teeth tips
  • Longueur:
    4.75"