Scissors, Disposable, Mortech
Scissors, Disposable, Mortech
Supplier:   Mopec
Scissors, Disposable, Mortech
Supplier:  Mopec
Certificates
Scissors, Disposable, Mortech
Scissors, Disposable, Mortech
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These disposable scissors are highly recommended for post-mortem infectious cases.
  • Straight cutting edge
  • Not autoclavable
  • Single use only
Package of 12. Available in multiple lengths and tips.
Product Family Options
Product Information
  • Longueur
    Blade Type
    Availability
    Price
  • 4.5"
    Sharp/Sharp
    Log in for availability
    Log in for availability
    Each
    Each - $64.72
    $64.72
    Each
       $64.72
    Supplier Number: AA601
  • 4.5"
    Sharp/Blunt
    Log in for availability
    Log in for availability
    Each
    Each - $64.72
    $64.72
    Each
       $64.72
    Supplier Number: AA602
  • 5.25"
    Sharp/Sharp
    Log in for availability
    Log in for availability
    Each
    Each - $104.64
    $104.64
    Each
       $104.64
    Supplier Number: AA603
  • 5.25"
    Sharp/Blunt
    Log in for availability
    Log in for availability
    Each
    Each - $104.64
    $104.64
    Each
       $104.64
    Supplier Number: AA604