Scissors, SuperCut Stevens Tenotomy, Mortech | Avantor
Scissors, SuperCut Stevens Tenotomy, Mortech
Supplier:   Mopec
Scissors, SuperCut Stevens Tenotomy, Mortech
Supplier:  Mopec
Certificates
Scissors, SuperCut Stevens Tenotomy, Mortech
Scissors, SuperCut Stevens Tenotomy, Mortech
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
SuperCut scissors are tempered and hardened for a longer, sharper life over ordinary steel. The SuperCut blade has a specially designed acute edge. They also feature rounded and polished finger rings. Helpful with delicate post-mortem dissection.
  • Premium grade stainless steel
  • Razor sharp cutting edge
  • Narrow sharp tips
Available with straight or curved edges.
Product Family Options
Informations sur le produit
  • Longueur
    Curvature
    Availability
    Price
  • 4.5"
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $498.73
    $498.73
    Each
       $498.73
    Supplier Number: AA535
  • 4.5"
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $586.11
    $586.11
    Each
       $586.11
    Supplier Number: AA536