Scissors, SuperCut Mayo, Mortech | Avantor
Scissors, SuperCut Mayo, Mortech
Supplier:   Mopec
Scissors, SuperCut Mayo, Mortech
Supplier:  Mopec
Certificates
Scissors, SuperCut Mayo, Mortech
Scissors, SuperCut Mayo, Mortech
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
SuperCut scissors are tempered and for a longer, sharper life over ordinary steel. The SuperCut blade has a specially designed razor sharp edge. The straight version can cut into surface area tissue while the curved version can cut into heavier or deeper tissue. They also feature rounded and polished finger rings.
  • Stainless steel
  • Slightly rounded tips
Available with straight or curved edges.
Product Family Options
Informations sur le produit
  • Longueur
    Curvature
    Availability
    Price
  • 5.5"
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $453.88
    $453.88
    Each
       $453.88
    Supplier Number: AA519
  • 6.75"
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $521.28
    $521.28
    Each
       $521.28
    Supplier Number: AA520
  • 9"
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $628.51
    $628.51
    Each
       $628.51
    Supplier Number: AA537
  • 5.5"
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $658.41
    $658.41
    Each
       $658.41
    Supplier Number: AA521
  • 6.75"
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $553.68
    $553.68
    Each
       $553.68
    Supplier Number: AA522
  • 9"
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $828.04
    $828.04
    Each
       $828.04
    Supplier Number: AA538