Scissors, SuperCut, Sharp/Blunt, Mortech
Scissors, SuperCut, Sharp/Blunt, Mortech
Supplier:   Mopec
Scissors, SuperCut, Sharp/Blunt, Mortech
Supplier:  Mopec
Certificates
Scissors, SuperCut, Sharp/Blunt, Mortech
Scissors, SuperCut, Sharp/Blunt, Mortech
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
SuperCut scissors are tempered and hardened for an extended, sharper life over ordinary steel. The SuperCut blade has a specially designed sharpened edge. They also feature rounded and polished finger rings. A general tool for cutting.
  • Stainless steel
  • Sharp/blunt tips
Available with straight or curved edges.
Product Family Options
Product Information
  • Longueur
    Curvature
    Availability
    Price
  • 5.5"
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $451.39
    $451.39
    Each
       $451.39
    Supplier Number: AA508
  • 5.5"
    Curved
    Log in for availability
    Log in for availability
    Each
    Each - $598.56
    $598.56
    Each
       $598.56
    Supplier Number: AA511